Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566084 Supplier Invitrogen™ Supplier No. PA566084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKSENCGVPED LLNGLKVTDT QEAECAGPPV PDPKNQHSQS KLLRDDEAHL QEDQGEEECF HDCSASFEEE P Highest antigen sequence identity to the following orthologs - mouse 71%, rat 74%.

This gene encodes a protein that belongs to the tetratrico peptide repeat superfamily of proteins. The encoded protein plays a role in protein-protein interactions, and binds to the Galpha subunit of G protein-coupled receptors to activate the Ras signaling pathway. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ttc1
Gene Accession No. Q99614
Gene Alias 4833412C19Rik; C79328; tetratricopeptide repeat domain 1; tetratricopeptide repeat protein 1; TPR repeat protein 1; TPR1; Ttc1
Gene Symbols Ttc1
Host Species Rabbit
Immunogen Recombinant Human TTC1. Recombinant protein control fragment (Product #RP-106778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7265
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.