Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC11 Antibody (27B4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TTC11 |
---|---|
Clone | 27B4 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
TTC11 Monoclonal antibody specifically detects TTC11 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
TTC11 | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CGI-135, Fis1, FIS1 homolog, fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae), fission 1 (mitochondrial outer membrane) homolog (yeast), H_NH0132A01.6, hFis1, mitochondrial fission 1 protein, tetratricopeptide repeat domain 11, Tetratricopeptide repeat protein 11, TTC11TPR repeat protein 11 | |
A synthesized peptide derived from Human TTC11 [UniProt Q9Y3D6] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
27B4 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Mitochondrial Fusion Proteins, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
51024 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title