Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584492 Supplier Invitrogen™ Supplier No. PA584492
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSDDPVVQQK AVLETEKRLL LMEEDQEEDE CRTTLNKTMI SPPQTAMKSA EEINSEAFLA SVEKDAKERA KRRRENKV Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 77%.

TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC12
Gene Accession No. Q9H892
Gene Alias E330017O07Rik; tetratricopeptide repeat domain 12; tetratricopeptide repeat protein 12; TPARM; TPR repeat protein 12; TTC12
Gene Symbols TTC12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC12. Recombinant protein control fragment (Product #RP-107057).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.