Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558431
Description
Immunogen sequence: NLLETLSKPD QIPLFYAGGI EILTEMINEC TEQTLFRMHN GFSIISDNEV IRRCFSTAGN DAVEEMVCVS VLKLWQAVCS RNEENQRVL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
TTC12 contains 3 TPR repeats. Hypermethylation of TTC12 gene may play a role in acute lymphoblastic leukemia. Haplotypic variants in DRD2, ANKK1, TTC12, and NCAM1 are associated with comorbid alcohol and drug dependence.
Specifications
TTC12 | |
Polyclonal | |
Unconjugated | |
TTC12 | |
E330017O07Rik; tetratricopeptide repeat domain 12; tetratricopeptide repeat protein 12; TPARM; TPR repeat protein 12; TTC12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H892 | |
TTC12 | |
Recombinant protein corresponding to Human TTC12. Recombinant protein control fragment (Product #RP-97355). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction