Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565668 Supplier Invitrogen™ Supplier No. PA565668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFESYKPEVQE LICVADRLGS LMQYETPGFL PNKRIHRAMG LAALEVMQAV QRTWTNSKVR MNGKTRLMQW RDMFDIAVKW RRIADPDQPV LWLDQMPARS L Highest antigen sequence identity to the following orthologs - mouse 96%, rat 98%.

TTC13 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC13
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC13
Gene Accession No. Q8NBP0
Gene Alias PSEC0076; tetratricopeptide repeat domain 13; tetratricopeptide repeat protein 13; TPR repeat protein 13; TTC13
Gene Symbols TTC13
Host Species Rabbit
Immunogen Recombinant Human TTC13. Recombinant protein control fragment (Product #RP-106343).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79573
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.