Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC14 Antibody (OTI9E11), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274655UV
Description
TTC14 Monoclonal specifically detects TTC14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TTC14 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TTC14 | |
Recombinant protein fragment corresponding to amino acids 429-770 of human TTC14 (NP_597719) produced in E.coli. | |
0.1 mL | |
151613.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI9E11 | |
Western Blot, Immunohistochemistry | |
DKFZp313M1015, DRDL5813, KIAA1980FLJ00166, PRO19630, tetratricopeptide repeat domain 14, tetratricopeptide repeat protein 14, TPR repeat protein 14 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction