Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC14 Monoclonal Antibody (OTI9E11)
GREENER_CHOICE

Catalog No. PIMA527055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527055 Supplier Invitrogen™ Supplier No. MA527055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTC14 Monoclonal Antibody for Western Blot, IHC (P)

TTC14 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTC14
Gene Accession No. Q96N46
Gene Alias 2700016E08Rik; 4930434D01Rik; 4931403I22Rik; 4933402I15Rik; AI662468; AU014779; AW561908; cI-44; DRDL5813; Kiaa1980; mKIAA1980; PRO19630; tetratricopeptide repeat domain 14; tetratricopeptide repeat protein 14; TPR repeat protein 14; Ttc14; UNQ5813; UNQ5813/PRO19630
Gene Symbols TTC14
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 429-770 of TTC14 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151613
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.