Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534887 Supplier Invitrogen™ Supplier No. PA534887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, K562, NCI-H929. Predicted reactivity: Mouse (95%), Rat (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TTC14 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC14
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene TTC14
Gene Accession No. Q96N46
Gene Alias 2700016E08Rik; 4930434D01Rik; 4931403I22Rik; 4933402I15Rik; AI662468; AU014779; AW561908; cI-44; DRDL5813; Kiaa1980; mKIAA1980; PRO19630; tetratricopeptide repeat domain 14; tetratricopeptide repeat protein 14; TPR repeat protein 14; Ttc14; UNQ5813; UNQ5813/PRO19630
Gene Symbols TTC14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TTC14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.