Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | TTC14 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TTC14 Polyclonal specifically detects TTC14 in Human samples. It is validated for Western Blot.Specifications
| TTC14 | |
| Polyclonal | |
| Rabbit | |
| Q96N46 | |
| 151613 | |
| Synthetic peptides corresponding to TTC14(tetratricopeptide repeat domain 14) The peptide sequence was selected from the N terminal of TTC14. Peptide sequence MDRDLLRQSLNCHGSSLLSLLRSEQQDNPHFRSLLGSAAEPARGPPPQHP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DKFZp313M1015, DRDL5813, KIAA1980FLJ00166, PRO19630, tetratricopeptide repeat domain 14, tetratricopeptide repeat protein 14, TPR repeat protein 14 | |
| TTC14 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title