Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557063
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557063 Supplier Invitrogen™ Supplier No. PA557063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFLHLGQNNF AEAHRFFTEI LRMDPRNAVA NNNAAVCLLY LGKLKDSLRQ LEAMVQQDPR HYLHESVLFN LTTMYELESS RSMQKKQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Component of the TRAPP complex, which is involved in endoplasmic reticulum to Golgi apparatus trafficking at a very early stage (PubMed:21525244, PubMed:28777934). Also plays a role in chromosome congression, kinetochore assembly and stability and controls the recruitment of CENPE to the kinetochores (PubMed:25918224). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRAPPC12
Gene Accession No. Q8WVT3
Gene Alias CGI-87; tetratricopeptide repeat domain 15; tetratricopeptide repeat protein 15; TPR repeat protein 15; Trafficking of membranes and mitosis; trafficking protein particle complex 12; trafficking protein particle complex subunit 12; TRAMM; TRAPPC12; TTC15; TTC-15
Gene Symbols TRAPPC12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC15. Recombinant protein control fragment (Product #RP-96199).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.