Learn More
Invitrogen™ TTC16 Polyclonal Antibody

Description
TTC16 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | TTC16 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TTC16 |
| Gene Accession No. | Q8NEE8 |
| Gene Alias | tetratricopeptide repeat domain 16; tetratricopeptide repeat protein 16; TPR repeat protein 16; TTC16 |
| Gene Symbols | TTC16 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.