Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584837
Description
Immunogen sequence: DLKGFKGDTP VTFIRAEFNQ VVLGDSAKIT VSPEGSAKYN FTSSFEFNPE GGITSDDLAH KPVFLTVTEV LPKEKKQKE.
TTC18 is a protein coding gene. Gene ontology (GO) annotation include cilium; extracellular exosome.
Specifications
TTC18 | |
Polyclonal | |
Unconjugated | |
CFAP70 | |
CFAP70; cilia and flagella associated protein 70; cilia- and flagella-associated protein 70; FLJ25765; tetratricopeptide repeat domain 18; tetratricopeptide repeat protein 18; tetratricopeptide repeat-containing protein (LOC118491); TPR repeat protein 18; TTC18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118491 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T0N1 | |
CFAP70 | |
Recombinant protein corresponding to Human CFAP70. Recombinant protein control fragment (Product #RP-108249). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction