Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558022
Description
Immunogen sequence: AVVDLLPLLE GQSSFQTTVP LHPVQGSPLE TPRSSAKQCS LEVKVLVAEP LLTTAQISGG NLLKVTLEAA YSVPESFIPT GPGQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 76%.
TTC18 is a protein coding gene. Gene ontology (GO) annotation include cilium; extracellular exosome.
Specifications
| TTC18 | |
| Polyclonal | |
| Unconjugated | |
| CFAP70 | |
| CFAP70; cilia and flagella associated protein 70; cilia- and flagella-associated protein 70; FLJ25765; tetratricopeptide repeat domain 18; tetratricopeptide repeat protein 18; tetratricopeptide repeat-containing protein (LOC118491); TPR repeat protein 18; TTC18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 118491 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T0N1 | |
| CFAP70 | |
| Recombinant protein corresponding to Human TTC18. Recombinant protein control fragment (Product #RP-97138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction