Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112295 Supplier Thermo Scientific Supplier No. PA5112295
Add to Cart
Add to Cart

TTC19 Polyclonal Antibody

TTC19 Polyclonal Antibody for Western Blot, ICC/IF, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC19
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene TTC19
Gene Accession No. Q6DKK2
Gene Alias 2010204O13Rik; 2810460C24Rik; AI505442; MC3DN2; RGD1311797; tetratricopeptide repeat domain 19; tetratricopeptide repeat protein 19, mitochondrial; tetratricopeptide repeat protein 19; tetratricopeptide repeat protein 19, mitochondrial; TPR repeat protein 19; TTC19
Gene Symbols TTC19
Host Species Rabbit
Immunogen E. coli-derived Human TTC19 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54902
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.