Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114786 Supplier Invitrogen™ Supplier No. PA5114786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TTC19.

This gene encodes a protein of unknown function. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TTC19
Gene Accession No. D4A6D7, Q6DKK2, Q8CC21
Gene Alias 2010204O13Rik; 2810460C24Rik; AI505442; MC3DN2; RGD1311797; tetratricopeptide repeat domain 19; tetratricopeptide repeat protein 19, mitochondrial; tetratricopeptide repeat protein 19; tetratricopeptide repeat protein 19, mitochondrial; TPR repeat protein 19; TTC19
Gene Symbols TTC19
Host Species Rabbit
Immunogen A synthesized peptide derived from human TTC19(Accession Q6DKK2), corresponding to amino acid residues A186-L236.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54902, 691506, 72795
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.