Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC21A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557301 Supplier Invitrogen™ Supplier No. PA557301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGAESNYMEK KELEQQGVST AEKLLREFYP HSDSSQTQLR LLQGLCRLAT REKANMEAAL GSFIQIAQAE KDSVPALLAL AQAYVFLKQI PK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC21A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC21A
Gene Accession No. Q8NDW8
Gene Alias IFT139A; STI2; Stress-inducible protein 2; testicular tissue protein Li 212; tetratricopeptide repeat domain 21A; tetratricopeptide repeat protein 21A; TPR domain containing STI2; TPR repeat protein 21A; TTC21A
Gene Symbols TTC21A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC21A. Recombinant protein control fragment (Product #RP-96431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.