Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC21B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TTC21B |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TTC21B Polyclonal specifically detects TTC21B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TTC21B | |
Polyclonal | |
Rabbit | |
Human | |
Q7Z4L5 | |
79809 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QRLLLQDSQNVEALRMQALYYVCREGDIEKASTKLENLGNALDAMEPQNAQLFYNITLAFSRTCGRSQLILQKIQTLLERAFSLNPQQSEFATELG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ATD4, JBTS11, KIAA1992, putative protein product of Nbla10696, tetratricopeptide repeat domain 21B, tetratricopeptide repeat protein 21B, THM1, TPR repeat protein 21B | |
TTC21B | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title