Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC21B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557293 Supplier Invitrogen™ Supplier No. PA557293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRLLLQDSQN VEALRMQALY YVCREGDIEK ASTKLENLGN ALDAMEPQNA QLFYNITLAF SRTCGRSQLI LQKIQTLLER AFSLNPQQSE FATELG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 72%.

The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. TTC21B (tetratricopeptide repeat domain 21B), also known as THM1, is a 1,316 amino acid protein that contains 19 TPR repeats and belongs to the TTC21 family. Localizing to cytoplasm and cytoskeleton, TTC21B exists as two alternatively spliced isoforms and is thought to negatively regulate Shh signal transduction. TTC21B may also be involved in retrograde intraflagellar transport in cilia, and is encoded by a gene that maps to human chromosome 2q24.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC21B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC21B
Gene Accession No. Q7Z4L5
Gene Alias 2410066K11Rik; aln; ATD4; IFT139; IFT139B; Intraflagellar transport 139 homolog; JBTS11; Kiaa1992; line 158; mKIAA1992; Nbla10696; NPHP12; putative protein product of Nbla10696; RGD1565122; SRTD4; tetratricopeptide repeat domain 21B; tetratricopeptide repeat protein 21B; Tetratricopeptide repeat-containing hedgehog modulator 1; Thm1; TPR repeat protein 21B; Ttc21b
Gene Symbols TTC21B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC21B. Recombinant protein control fragment (Product #RP-102668).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.