Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC21B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981919
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1919 Supplier Bioss Supplier No. BS12069R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen TTC21B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TTC21B
Gene Accession No. Q7Z4L5
Gene Alias ATD4; IFT139; IFT139B; Intraflagellar transport 139 homolog; JBTS11; KIAA1992; Nbla10696; NPHP12; putative protein product of Nbla10696; SRTD4; tetratricopeptide repeat domain 21B; Tetratricopeptide repeat protein 21B; THM1; TPR repeat protein 21B; TTC21B
Gene Symbols TTC21B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TTC21B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79809
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.