Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571278 Supplier Invitrogen™ Supplier No. PA571278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 86%; Rabbit: 86%; Rat: 100%.

This gene encodes a protein with seven tetratricopeptide (TPR) repeats. Tetratricopeptide repeat containing motifs are found in a variety of proteins and may mediate protein-protein interactions and chaperone activity. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TTC22
Gene Accession No. Q5TAA0
Gene Alias 4732467L16Rik; LOW QUALITY PROTEIN: tetratricopeptide repeat protein 22; RGD1561518; tetratricopeptide repeat domain 22; tetratricopeptide repeat protein 22; TPR repeat protein 22; TTC22
Gene Symbols TTC22
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human TTC22.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55001
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.