Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557142 Supplier Invitrogen™ Supplier No. PA557142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMPDRNHLAC AKADLEEVVR VCPGFKAYLD IGQVYYYMGV DAVQELLAVD EAALNQALVF LAKAGESELG ATLPELQLLR GKCLRIKGED Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

This gene encodes a protein with seven tetratricopeptide (TPR) repeats. Tetratricopeptide repeat containing motifs are found in a variety of proteins and may mediate protein-protein interactions and chaperone activity. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC22
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC22
Gene Accession No. Q5TAA0
Gene Alias 4732467L16Rik; LOW QUALITY PROTEIN: tetratricopeptide repeat protein 22; RGD1561518; tetratricopeptide repeat domain 22; tetratricopeptide repeat protein 22; TPR repeat protein 22; TTC22
Gene Symbols TTC22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC22. Recombinant protein control fragment (Product #RP-93975).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55001
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.