Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599304 Supplier Invitrogen™ Supplier No. PA599304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TTC23.

Participates positively in the ciliary Hedgehog (Hh) signaling. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC23
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TTC23
Gene Accession No. Q5W5X9, Q8CHY7
Gene Alias 1600012K10Rik; Cervical cancer proto-oncogene 8 protein; HCC8; HCC-8; RGD1310409; tetratricopeptide repeat domain 23; tetratricopeptide repeat protein 23; TPR repeat protein 23; TTC23
Gene Symbols TTC23
Host Species Rabbit
Immunogen A synthesized peptide derived from human TTC23(Accession Q5W5X9), corresponding to amino acid residues I177-C227.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64927, 67009
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.