Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC23 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986926
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6926 Supplier Bioss Supplier No. BS5947R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The tetratricopeptide repeat (TPR) is a structural motif. It consists in a degenerate 34 amino acid sequence motif identified in a wide variety of proteins. It is found in tandem arrays of 3-16 motifs, which form scaffolds to mediate protein-protein interactions and often the assembly of multiprotein complexes. These repeats usually fold together to produce a single, linearsolenoid domain called TPR domain.

Specifications

Antigen TTC23
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TTC23
Gene Accession No. Q5W5X9
Gene Alias 1600012K10Rik; Cervical cancer proto-oncogene 8 protein; HCC8; HCC-8; tetratricopeptide repeat domain 23; Tetratricopeptide repeat protein 23; TPR repeat protein 23; TTC23
Gene Symbols TTC23
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TTC23.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64927
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.