Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC23L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569870 Supplier Invitrogen™ Supplier No. PA569870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Horse: 86%; Human: 100%; Rabbit: 79%.

TTC23L (Tetratricopeptide Repeat Domain 23 Like) is a Protein Coding gene. An important paralog of this gene isTTC23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC23L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TTC23L
Gene Accession No. Q6PF05
Gene Alias 4930401A09Rik; MC25-1; tetratricopeptide repeat domain 23 like; tetratricopeptide repeat domain 23-like; tetratricopeptide repeat protein 23-like; Ttc23l
Gene Symbols TTC23L
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human TTC23L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153657
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.