Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC23L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571558
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571558 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571558 Supplier Invitrogen™ Supplier No. PA571558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC23L Polyclonal Antibody for Western Blot, IHC (P)

TTC23L (Tetratricopeptide Repeat Domain 23 Like) is a Protein Coding gene. An important paralog of this gene isTTC23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC23L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TTC23L
Gene Accession No. A6H6E9, Q6PF05
Gene Alias 4930401A09Rik; MC25-1; tetratricopeptide repeat domain 23 like; tetratricopeptide repeat domain 23-like; tetratricopeptide repeat protein 23-like; Ttc23l
Gene Symbols TTC23L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 319-348 amino acids from the C-terminal region of human TTC23L.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153657, 75777
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.