Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570945 Supplier Invitrogen™ Supplier No. PA570945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a tetratricopeptide repeat domain-containing protein that localizes to ciliary axonmenes and plays a role in the docking of the outer dynein arm to cilia. Mutations in this gene cause severely reduced ciliary motility and the disorder CILD35 (ciliary dyskinesia, primary, 35). Primary ciliary dyskinesia is often associated with recurrent respiratory infections, immotile spermatozoa, and situs inversus; an inversion in left-right body symmetry. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TTC25
Gene Accession No. Q96NG3
Gene Alias tetratricopeptide repeat domain 25; tetratricopeptide repeat protein 25; TPR repeat protein 25; TTC25
Gene Symbols TTC25
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human TTC25.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83538
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.