Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598219
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598219 Supplier Invitrogen™ Supplier No. PA598219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTC26 Polyclonal Antibody for Western Blot, ELISA

Component of the intraflagellar transport (IFT) complex B required for transport of proteins in the motile cilium. Required for transport of specific ciliary cargo proteins related to motility, while it is neither required for IFT complex B assembly or motion nor for cilium assembly. Required for efficient coupling between the accumulation of GLI2 and GLI3 at the ciliary tips and their dissociation from the negative regulator SUFU. Plays a key role in maintaining the integrity of the IFT complex B and the proper ciliary localization of the IFT complex B components. Not required for IFT complex A ciliary localization or function. Essential for maintaining proper microtubule organization within the ciliary axoneme. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC26
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TTC26
Gene Accession No. A0AVF1, Q8BS45
Gene Alias 9330141E21Rik; 9430097H08Rik; DYF13; dyf-13; Hop; Ift56; intraflagellar transport protein 56; Protein hop-sterile; tetratricopeptide repeat domain 26; tetratricopeptide repeat protein 26; TPR repeat protein 26; Ttc26
Gene Symbols TTC26
Host Species Rabbit
Immunogen Recombinant Human Intraflagellar transport protein 56 protein (5-122AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 264134, 79989
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.