Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554942 Supplier Invitrogen™ Supplier No. PA554942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DCPHVDDCVF AAEFMSNDYV RVTQLYCDGV GVQYKDYIQS ERNLEFDICS IWCSKPISVL QDYCDAIKIN IFWPLLFQHQ NSSVISRLHP CVDANNSRAS EINLKKLQHL ELMEDIVDLA KKVANDS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

E3 ubiquitin-protein ligase which catalyzes the formation of 'Lys-48'-polyubiquitin chains (PubMed:20059950, PubMed:30696809). Mediates the ubiquitination and subsequent degradation of phosphorylated Akt (AKT1, AKT2 and AKT3) in the nucleus (PubMed:20059950). Acts as a terminal regulator of Akt signaling after activation; its phosphorylation by Akt, which is a prerequisite for ubiquitin ligase activity, suggests the existence of a regulation mechanism required to control Akt levels after activation (PubMed:20059950). Positively regulates TGFB1-induced epithelial-mesenchymal transition and myofibroblast differentiation by mediating the ubiquitination and subsequent degradation of SMURF2 (PubMed:30696809). Regulates neuronal differentiation by regulating actin remodeling and Golgi organization via a signaling cascade involving RHOA, CIT and ROCK (PubMed:17488780, PubMed:24695496). Inhibits cell proliferation (PubMed:30203323). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC3
Gene Accession No. P53804
Gene Alias DCRR1; E3 ubiquitin-protein ligase TTC3; Protein DCRR1; RING finger protein 105; RING-type E3 ubiquitin transferase TTC3; RNF105; tetratricopeptide repeat domain 3; tetratricopeptide repeat protein 3; TPR repeat protein 3; TPR repeat protein D; TPRD; TPRDIII; TTC3
Gene Symbols TTC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC3. Recombinant protein control fragment (Product #RP-91795).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.