Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC30A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562465
Description
Immunogen sequence: FRKSVEFCND HDVWKLNVAH VLFMQENKYK EAIGFYEPIV KKHYDNILNV SAIVLANLCV SYIMTSQNEE AEELMRKIEK EEEQLSYDDP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

Specifications
TTC30A | |
Polyclonal | |
Unconjugated | |
TTC30A | |
IFT70A; tetratricopeptide repeat domain 30A; tetratricopeptide repeat domain 30B; tetratricopeptide repeat protein 30A; TPR repeat protein 30A; TTC30A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92104 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WT1 | |
TTC30A | |
Recombinant protein corresponding to Human TTC30A. Recombinant protein control fragment (Product #RP-104467). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction