Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC30B Polyclonal Antibody

Supplier: Thermo Scientific PA5113127
Description
Specifications
| TTC30B | |
| Polyclonal | |
| Unconjugated | |
| TTC30B | |
| 2510042P03Rik; fleer; IFT70; IFT70B; novel tetratrico peptide repeat (TPR) containing protein; Tcc30b; tetratricopeptide repeat domain 30B; tetratricopeptide repeat protein 30B; TPR repeat protein 30B; TTC30B | |
| Rabbit | |
| Protein G | |
| RUO | |
| 150737 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N4P2 | |
| TTC30B | |
| Recombinant Human Tetratricopeptide repeat protein 30B protein (261-600AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction