Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC32 Antibody (OTI1F7), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274656MFV500
Description
TTC32 Monoclonal antibody specifically detects TTC32 in Human samples. It is validated for Flow CytometrySpecifications
TTC32 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1F7 | |
Flow Cytometry | |
tetratricopeptide repeat domain 32, tetratricopeptide repeat protein 32, TPR repeat protein 32 | |
Full length human recombinant protein of human TTC32(NP_001008238) produced in HEK293T cell. | |
0.1 mL | |
Zinc Finger | |
130502 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction