Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC32 Monoclonal Antibody (OTI2B9), TrueMAB™, OriGene
SDP

Catalog No. 502542364
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2364 30 μL
50-167-2488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2364 Supplier OriGene Supplier No. TA501343S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTC32 Monoclonal antibody specifically detects TTC32 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TTC32
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B9
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ttc32
Gene Accession No. Q5I0X7
Gene Alias 1700013G20Rik; tetratricopeptide repeat domain 32; tetratricopeptide repeat protein 32; TPR repeat protein 32; TTC32
Host Species Mouse
Immunogen Full length human recombinant protein of human TTC32 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130502
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.