Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC32 Antibody (OTI1F7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274656PE
Description
TTC32 Monoclonal specifically detects TTC32 in Human samples. It is validated for Flow Cytometry.Specifications
TTC32 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
TTC32 | |
Full length recombinant protein of human TTC32 (NP_001008238) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI1F7 | |
Flow Cytometry | |
tetratricopeptide repeat domain 32, tetratricopeptide repeat protein 32, TPR repeat protein 32 | |
Mouse | |
Affinity Purified | |
Zinc Finger | |
130502.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction