Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC33 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299640AF405
Description
TTC33 Polyclonal specifically detects TTC33 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TTC33 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human TTC33. | |
0.1 mL | |
Signal Transduction | |
23548 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction