Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC33 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237758H
Description
TTC33 Polyclonal antibody specifically detects TTC33 in Human, Mouse samples. It is validated for Western BlotSpecifications
TTC33 | |
Polyclonal | |
Western Blot | |
osmosis responsive factor, Osmosis-responsive factor, OSRF, tetratricopeptide repeat domain 33, tetratricopeptide repeat protein 33, TPR repeat protein 33 | |
The immunogen this antibody was made to, maps to a region between residue 212 to 262 of human Tetratricopeptide Repeat Domain 33 using the numbering given in entry NP_036514.1 (GeneID 23548). | |
0.1 mL | |
Signal Transduction | |
23548 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction