Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC34 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA564291

Catalog No. PIPA564291


Only null left
Add to Cart

Description

Description

Immunogen sequence: TCLAELQEFG RALRDLDHVL QEALGDGDLP RRAEDFCRQG RLLLSLGDEA Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TTC34
Polyclonal
Unconjugated
TTC34
tetratricopeptide repeat domain 34; tetratricopeptide repeat protein 34; TPR repeat protein 34; TPR repeat-containing protein ENSP00000383873; TTC34
Rabbit
Antigen affinity chromatography
RUO
100287898
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A8MYJ7
TTC34
Recombinant protein corresponding to Human TTC34. Recombinant protein control fragment (Product #RP-104177).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.