Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543159 Supplier Invitrogen™ Supplier No. PA543159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IQLYDRILQE DPTNTAARKR KIAIRKAQGK NVEAIRELNE YLEQFVGDQE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The exact function of TTC35 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC35
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Emc2
Gene Accession No. Q15006
Gene Alias 4921531G14Rik; AV060620; AW209495; emc2; ER membrane protein complex subunit 2; KIAA0103; KIAA0103-like protein; RGD1310430; tetratricopeptide repeat domain 35; tetratricopeptide repeat protein 35; Tetratricopeptide repeat protein 35-like protein; TPR repeat protein 35; TTC35; Unknown (protein for MGC:128301); zgc:73154; zgc:86891
Gene Symbols Emc2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TTC35.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9694
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.