Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554617 Supplier Invitrogen™ Supplier No. PA554617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELYDVTWEEM RDKMRKWREE NSRNSEQIVE VGEELINEYA SKLGDDIWII YEQVMIAALD YGRDDLALFC LQELRRQFPG SH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 73%.

The exact function of TTC35 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC35
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Emc2
Gene Accession No. Q15006
Gene Alias 4921531G14Rik; AV060620; AW209495; emc2; ER membrane protein complex subunit 2; KIAA0103; KIAA0103-like protein; RGD1310430; tetratricopeptide repeat domain 35; tetratricopeptide repeat protein 35; Tetratricopeptide repeat protein 35-like protein; TPR repeat protein 35; TTC35; Unknown (protein for MGC:128301); zgc:73154; zgc:86891
Gene Symbols Emc2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC35. Recombinant protein control fragment (Product #RP-93768).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.