Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC39A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565358 Supplier Invitrogen™ Supplier No. PA565358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLTNQFSEALS YLKPRTKESM YHSLTYATIL EMQAMMTFDP QDILLAGNMM KEAQMLCQRH RRKSSVTDSF SSLVNRPTLG QFTEEEIHAE V Highest antigen sequence identity to the following orthologs - mouse 93%, rat 96%.

TTC39A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC39A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC39A
Gene Accession No. Q5SRH9
Gene Alias 4922503N01Rik; C1orf34; DEME-6; Differentially expressed in MCF7 with estradiol protein 6; KIAA0452; RGD1311126; testicular tissue protein Li 213; tetratricopeptide repeat domain 39A; tetratricopeptide repeat protein 39A; TPR repeat protein 39A; Ttc39a
Gene Symbols TTC39A
Host Species Rabbit
Immunogen Recombinant Human TTC39A. Recombinant protein control fragment (Product #RP-104772).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.