Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC39B Rabbit anti-Mouse, Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS211925UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2119-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2119-25UG Supplier MilliporeSigma Supplier No. ABS211925UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-TTC39B, Cat. No. ABS2119, is a rabbit polyclonal antibody that detects Tetratricopeptide repeat protein 39B and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC39B
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.
Gene Alias Tetratricopeptide repeat protein 39B;TPR repeat protein 39B
Gene Symbols TTC39B;C9orf52
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human Tetratricopeptide repeat protein (TTC39B).
Molecular Weight of Antigen ∽90 kDa observed; 76.96 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects TTC39B in human and murine cells. It targets an epitope with in 20 amino acids from the C-terminal half.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.