Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTC4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TTC4 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TTC4 Polyclonal specifically detects TTC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TTC4 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7268 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AQSDFISAFHEDSRFIDHLMVMFGETPSWDLEQKYCPDNLEVYFEDEDRAELYRVPAKSTLLQVLQHQRYFVKALTPAFLVCVGSSPFCKNFLRGRKVYQI | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
DKFZp781B0622, FLJ41930, MGC5097, tetratricopeptide repeat domain 4, tetratricopeptide repeat protein 4, TPR repeat protein 4 | |
TTC4 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title