Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559647 Supplier Invitrogen™ Supplier No. PA559647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQSDFISAFH EDSRFIDHLM VMFGETPSWD LEQKYCPDNL EVYFEDEDRA ELYRVPAKST LLQVLQHQRY FVKALTPAFL VCVGSSPFCK NFLRGRKVYQ I Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

Effector protein for Rho-type GTPases. Regulates actin filament reorganization via its interaction with the Arp2/3 complex. Important for efficient actin polymerization. Possible regulator of lymphocyte and platelet function. Mediates actin filament reorganization and the formation of actin pedestals upon infection by pathogenic bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ttc4
Gene Accession No. O95801
Gene Alias 2810002P21Rik; AF007005; L62; My044; tetratricopeptide repeat domain 4; tetratricopeptide repeat protein 4; TPR repeat protein 4; TTC4
Gene Symbols Ttc4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC4. Recombinant protein control fragment (Product #RP-99848).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7268
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.