Learn More
TTC4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305600AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The 34-amino acid tetratricopeptide repeat (TPR) motif is found in a variety of proteins and may mediate protein-protein or protein-membrane interactions.[supplied by OMIM].Specifications
TTC4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95801 | |
TTC4 | |
Between 225 and 275 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
TTC4 | |
TPR repeat protein 4, tetratricopeptide repeat protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7268 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.