Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558570 Supplier Invitrogen™ Supplier No. PA558570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEEKVKEPKQ SQSPAPIKQL EDLPMSIEEW ASYSCPEEVL SVLKQDRSDS TVNPSSIQKP TYSLYFLDHL Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 31%.

The TTC40 gene, which stands for Tetratricopeptide repeat domain 40, is a member of the TPR protein family characterized by highly conserved motifs that facilitate protein-protein interactions. This gene plays a significant role in various cellular processes, including signal transduction and protein assembly. TTC40 has been identified as a novel methylated gene in nasopharyngeal carcinoma, where its expression is significantly reduced in tumor tissues due to hypermethylation. This epigenetic alteration suggests a potential role of TTC40 in tumorigenesis, possibly acting as a tumor suppressor. The decreased expression of TTC40 in cancerous tissues highlights its importance in maintaining cellular homeostasis and integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC40
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP46
Gene Accession No. Q8IYW2
Gene Alias 9330101J02Rik; bA288G11.4; bA288G11.5; bB137A17.2; bB137A17.3; C10orf123; C10orf124; C10orf92; C10orf93; CFAP46; cilia and flagella associated protein 46; cilia- and flagella-associated protein 46; E030019B06Rik; Gm170; protein CFAP46; RGD1559985; similar to chromosome 10 open reading frame 92; tetratricopeptide repeat domain 40; tetratricopeptide repeat protein 40; TPR repeat-containing protein C10orf93; TTC40
Gene Symbols CFAP46
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC40. Recombinant protein control fragment (Product #RP-103937).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.