Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589101 Supplier Invitrogen™ Supplier No. PA589101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMADEEEEVK PILQKLQELV DQLYSFRDCY FETHSVEDAG RKQQDVQKEM EKTLQQMEEV VGSVQGKAQV LMLTGKALNV TPDYSPKAEE LLSKAVKLEP ELVEAWNQLG EVYWKKGDVA AAHTCFSGAL THCRNKVSLQ NLSMVLRQLR TDTEDEHSHH VMDSVRQAKL AVQMDVHDGR SWYILGNSYL SLYFSTGQNP KISQQALSAY AQAEKVDRKA SSNPDLHLNR ATLHKYEESY GEALEGFSRA AALDPAWPEP RQREQQLLEF LDRLTSLLES KGKVKTKKLQ SMLGSLRPAH; Positive Samples: A-431, MCF-7, Mouse spleen, Mouse testis, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Tetratricopeptide repeat protein 5 (TTC5) is a member of a diverse group of functionally distinct proteins that are characterized by containing one or more tetratricopeptide repeats. Each motif consists of two anti-parallel a-helices such that tandem arrays of TPR motifs generate a right-handed helical structure with an amphipathic channel that may serve to accommodate the complementary region of a target protein. While the exact function of TTC5 remains unclear, it is thought that the TPR motifs serve to mediate protein-protein interactions such as those seen with protein chaperones HSP70 and HSP90 and some proteins involved in cell stress response signaling pathways such as protein phosphatase 5, suggesting that TTC5 may also function via protein-protein interactions mediated by its TPR motifs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TTC5
Gene Accession No. Q8N0Z6, Q99LG4
Gene Alias Strap; stress-responsive activator of p300; tetratricopeptide repeat domain 5; tetratricopeptide repeat protein 5; TPR repeat protein 5; TTC5
Gene Symbols TTC5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TTC5 (NP_6123852).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219022, 91875
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.