Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC7A Mouse anti-Human, Rat, Clone: OTI6D2, liquid, TrueMAB™
SDP

Catalog No. 501684512
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4512 Supplier OriGene Supplier No. TA812308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen TTC7A
Applications Western Blot
Classification Monoclonal
Clone OTI6D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TTC7A
Gene Accession No. Q9ULT0
Gene Alias GIDID, MINAT, TTC7
Gene Symbols TTC7A
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-450 of human TTC7A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362696, 57217
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.