Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTC7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557813 Supplier Thermo Scientific Supplier No. PA557813
Add to Cart
Add to Cart

TTC7A Polyclonal Antibody

TTC7A Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC7A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTC7A
Gene Accession No. Q9ULT0
Gene Alias 1110035E02Rik; 1700007L07Rik; fsn; GIDID; hea; KIAA1140; MINAT; tetratricopeptide repeat domain 7; tetratricopeptide repeat domain 7A; tetratricopeptide repeat protein 7A; TPR repeat protein 7A; Ttc7; TTC7A
Gene Symbols TTC7A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC7A. Recombinant protein control fragment (Product #RP-95973).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57217
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.