Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTC7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563561 Supplier Invitrogen™ Supplier No. PA563561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SECQWERIPE LVKQLSAKLI ANDDMAELLL GESKLEQYLK EHPLRQGASP RGPKPQLTEV RKHLTAALDR GNLKSEFLQE SNLIMAKLNY VEGDY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 35%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TTC7B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ttc7b
Gene Accession No. Q86TV6
Gene Alias AA408451; c14_5685; tetratricopeptide repeat domain 7 like 1; tetratricopeptide repeat domain 7B; Tetratricopeptide repeat protein 7B; tetratricopeptide repeat protein 7-like-1; TPR repeat protein 7B; TPR repeat protein 7-like-1; Ttc7b; TTC7L1
Gene Symbols Ttc7b
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTC7B. Recombinant protein control fragment (Product #RP-101938).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145567
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.