Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563750
Description
Immunogen sequence: HYHDALNIID MALSEYPENF ILLFSKVKLQ SLCRGPDEAL LTCKHMLQIW KSCYNLTNPS DSGRGSSLLD RTIADRRQLN TITLPDFSD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

Specifications
TTC7B | |
Polyclonal | |
Unconjugated | |
Ttc7b | |
AA408451; c14_5685; tetratricopeptide repeat domain 7 like 1; tetratricopeptide repeat domain 7B; Tetratricopeptide repeat protein 7B; tetratricopeptide repeat protein 7-like-1; TPR repeat protein 7B; TPR repeat protein 7-like-1; Ttc7b; TTC7L1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
145567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TV6 | |
Ttc7b | |
Recombinant protein corresponding to Human TTC7B. Recombinant protein control fragment (Product #RP-101939). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction