Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTC9B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559999
Description
Immunogen sequence: HGSARPGPTP EPSGSLGAAL DSSLRAAVAF KAEGQRCYRE KKFREAI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

Specifications
TTC9B | |
Polyclonal | |
Unconjugated | |
TTC9B | |
2900074C18Rik; tetratricopeptide repeat domain 9B; tetratricopeptide repeat protein 9B; TPR repeat protein 9B; Ttc9b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148014 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6N2 | |
TTC9B | |
Recombinant protein corresponding to Human TTC9B. Recombinant protein control fragment (Product #RP-99176). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction